Learn More
Enzo Life Sciences Pyocyanin (1µmol). CAS: 85-66-5
SDP
Supplier:  Enzo Life Sciences ENZ53001C001
Encompass_Preferred
Cytotoxic pigment secreted by Pseudomonas aeruginosa, which frequently infects the lungs of immuno-suppressed patients, as well as those with cystic fibrosis. Pyocyanin toxicity results from its ability to undergo reduction by NAD(P)H and subsequent generation of superoxide and H2O2 directly in the lungs. Alternative name: Sanasin, 1-hydroxy-N-methylphenazine. Formulation: Lyophilized. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -80°C.
Catalog No. 50-201-2647