Learn More
Cayman Chemical Pyocyanin, 85-66-5, 5 mg
SDP
Supplier:  Cayman Chemical 100095945
Encompass_Preferred
Activates the aryl hydrocarbon receptor (Ki = 5.4 µM), inducing the expression of both detoxifying enzymes, resulting in pyocyanin degradation, and cytokines that facilitate the clearance of bacteria; accelerates neutrophil apoptosis, resulting in resolution of acute inflammation
Catalog No. NC1500788