Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PyOxim, 97%, Thermo Scientific™

Catalog No. AC441410050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441410050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441410050 Supplier Thermo Scientific Chemicals Supplier No. 441410050
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 153433-21-7
Molecular Formula C17H29F6N5O3P2
Molecular Weight (g/mol) 527.39
MDL Number MFCD18643381
InChI Key RDWDVLFMPFUBDV-UHFFFAOYSA-N
Synonym pyoxim, pyoxime, ethyl cyano hydroxyimino acetato-tri-1-pyrrolidinyl-phosphonium hexafluorophosphate, cyano-ethoxycarbonyl methyleneaminooxy tris 1-pyrrolidinyl phosphonium hexafluorophosphate
PubChem CID 46910530
IUPAC Name [(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NO[P+](N1CCCC1)(N1CCCC1)N1CCCC1)C#N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 527.39
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material PyOxim
Hazard Statement GHS H Statement
Risk of explosion if heated under confinement.
Precautionary Statement GHS P Statement
Do not get in eyes,on skin,or on clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.