Learn More
Key Scientific Products PYR
SDP
Catalog No. NC0636045
Encompass_Preferred
The Pyrrolidonyl Arylamidase (PYR) test is a rapid test which is used for the presumptive identification of group A beta-hemolytic Streptococci and Enterococci. TheEnzymatic hydrolysis of PYR (L-pyroglutamic acid �-naphthylamide, aka pyrrolydonyl-�-naphthylamide) releases free �-naphthylamine which is detected and shown by the color change after adding PEP reagent.
Catalog No. NC0636045 Supplier Key Scientific Products Supplier No. K1538B
The Pyrrolidonyl Arylamidase (PYR) test is a rapid test which is used for the presumptive identification of group A beta-hemolytic Streptococci and Enterococci. TheEnzymatic hydrolysis of PYR (L-pyroglutamic acid �-naphthylamide, aka pyrrolydonyl-�-naphthylamide) releases free �-naphthylamine which is detected and shown by the color change after adding PEP reagent.