Learn More
Medchemexpress LLC HY-13296 10mg , PYR-41 CAS:418805-02-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1329610MG
Encompass_Preferred
HY-13296 10mg PYR-41 CAS: 418805-02-4 PYR-41 is a selective and cell permeable inhibitor of ubiquitin-activating enzyme E1 with an IC50 of < 10 μM, with little activity at E2 and E3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0053