Learn More
Selleck Chemical LLC PYR-41
SDP
Supplier:  Selleck Chemical LLC S71291G
Encompass_Preferred
PYR-41 is the first cell-permeable inhibitor of ubiquitin-activating enzyme E1 with no activity at E2 PYR-41 induce apoptosis
Catalog No. 50-000-01027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.