Learn More
Abcam PYR-41, Ubiquitin-activating enzyme (E1) inhibitor, 25MG
Catalog No. 502706588
Encompass_Preferred
MW 371.3 Da, Purity >98%. Ubiquitin-activating enzyme (E1) inhibitor (IC₅₀ = 5 μM). Inhibits ubiquitination and proteasomal degradation. Additionally increases sumoylation of proteins and induces p53-mediated apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6588 Supplier Abcam Supplier No. AB14146925MG
MW 371.3 Da, Purity >98%. Ubiquitin-activating enzyme (E1) inhibitor (IC₅₀ = 5 μM). Inhibits ubiquitination and proteasomal degradation. Additionally increases sumoylation of proteins and induces p53-mediated apoptosis.
The product is subject to the following: Abcam Restricted Use Statement