Learn More
Abcam PYR-41, Ubiquitin-activating enzyme (E1) inhibitor, 5MG
Catalog No. 502706587
Encompass_Preferred
MW 371.3 Da, Purity >98%. Ubiquitin-activating enzyme (E1) inhibitor (IC₅₀ = 5 μM). Inhibits ubiquitination and proteasomal degradation. Additionally increases sumoylation of proteins and induces p53-mediated apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6587 Supplier Abcam Supplier No. AB1414695MG
MW 371.3 Da, Purity >98%. Ubiquitin-activating enzyme (E1) inhibitor (IC₅₀ = 5 μM). Inhibits ubiquitination and proteasomal degradation. Additionally increases sumoylation of proteins and induces p53-mediated apoptosis.
The product is subject to the following: Abcam Restricted Use Statement