Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyr-Arg-Thr-Lys-Arg-AMC TFA | 1255501-99-5 | 99.5% | 941.95 | 10 MG
SDP

Catalog No. 5000397929
Encompass_Preferred

Pyr-Arg-Thr-Lys-Arg-AMC TFA is an AMC peptide. This peptide is specifically hydrolyzed by proteases such as trypsin and thrombin, making it valuable for determining protease activity and evaluating the potency of enzyme inhibitors.

  • Specifically hydrolyzed by proteases such as trypsin and thrombin
  • Useful for determining protease activity
  • Useful for determining the potency of enzyme inhibitors
  • Targets thrombin
  • Involves metabolic enzyme/protease pathway
  • Appearance: solid, white to off-white
  • Soluble in H₂O at 100 mg/mL (106.16 mM; requires ultrasonic)

Catalog No. 50-003-97929 Supplier Medchemexpress LLC Supplier No. HYP4349A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyr-Arg-Thr-Lys-Arg-AMC TFA is an AMC peptide. This peptide is specifically hydrolyzed by proteases such as trypsin and thrombin, making it valuable for determining protease activity and evaluating the potency of enzyme inhibitors.

  • Specifically hydrolyzed by proteases such as trypsin and thrombin
  • Useful for determining protease activity
  • Useful for determining the potency of enzyme inhibitors
  • Targets thrombin
  • Involves metabolic enzyme/protease pathway
  • Appearance: solid, white to off-white
  • Soluble in H₂O at 100 mg/mL (106.16 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.