Learn More
Apexbio Technology LLC Pyrophosphatase Inorganic (E. coli) 10 units
SDP
Catalog No. 503107130
Encompass_Preferred
Pyrophosphatase Inorganic (E coli) purified from recombinant E coli strain carrying the E coli inorganic pyrophosphatase gene Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme that catalyzes the hydrolysis of inorganic pyrophosphate into orthophosphate
Catalog No. 50-310-7130 Supplier Apexbio Technology LLC Supplier No. K108710
Pyrophosphatase Inorganic (E coli) purified from recombinant E coli strain carrying the E coli inorganic pyrophosphatase gene Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme that catalyzes the hydrolysis of inorganic pyrophosphate into orthophosphate