Learn More
TARGETMOL CHEMICALS INC PYR3 5MG
Supplier: TARGETMOL CHEMICALS INC T166875MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pyr3 is a selective transient receptor potential canonical channel 3 inhibitor. Pyr3 inhibits TRPC3-mediated Ca2+ influx in a dose-dependent manner(IC50 = 700 nM). purity: 99%