Learn More
Medchemexpress LLC HY-108465 5mg Medchemexpress, Pyr3 CAS:1160514-60-2 Purity:>98%
SDP
Catalog No. 501968553
Encompass_Preferred
Medchemexpress, HY-108465 5mg Pyr3 CAS:1160514-60-2 Pyr3 is a selective inhibitor of transient receptor potential canonical channel 3 (TRPC3), with an IC50 of 700 nM for TRPC3-mediated Ca2+ influx. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8553 Supplier Medchemexpress LLC Supplier No. HY1084655MG
Medchemexpress, HY-108465 5mg Pyr3 CAS:1160514-60-2 Pyr3 is a selective inhibitor of transient receptor potential canonical channel 3 (TRPC3), with an IC50 of 700 nM for TRPC3-mediated Ca2+ influx. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.