Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Pyr3)-Amyloid β-Protein (3-42) (TFA) | 99.6% | 4309.86 | 1 MG
SDP

Catalog No. 5000398220
Encompass_Preferred

(Pyr3)-Amyloid β-Protein (3-42) TFA is a predominant amyloid β-peptide structure found in the human brain of Alzheimer's disease and Down's syndrome patients. It is suggested to accumulate in the brain and trigger the formation of insoluble amyloid β-peptide deposits. This product is for research use only.

  • Molecular weight: 4309.86 (free acid)
  • Appears as a solid
  • Color is white to off-white

Catalog No. 50-003-98220 Supplier Medchemexpress LLC Supplier No. HYP4882A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Pyr3)-Amyloid β-Protein (3-42) TFA is a predominant amyloid β-peptide structure found in the human brain of Alzheimer's disease and Down's syndrome patients. It is suggested to accumulate in the brain and trigger the formation of insoluble amyloid β-peptide deposits. This product is for research use only.

  • Molecular weight: 4309.86 (free acid)
  • Appears as a solid
  • Color is white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.