Learn More
Medchemexpress LLC Pyraclonil | 158353-15-2 | MFCD11112213 | 97.0% | 314.77 g/mol | C15H15ClN6 | 10 MG
SDP
Catalog No. 502081977
Encompass_Preferred
Pyraclonil is a proporphyrinogen oxidase (PPO) inhibitor and herbicide agent supplied as a research-grade solid. The compound has molecular formula C15H15ClN6, molecular weight 314.77 g/mol, and supplier-reported purity of approximately 97%. Intended for laboratory research use; consult the safety data sheet (SDS) for handling and storage instructions.
  • PPO inhibitor suitable for herbicide and biochemical research.
  • High purity (~97%) appropriate for analytical and biological assays.
  • Supplied in a small research pack for method development and screening.
  • Defined molecular weight and formula support accurate dosing.
  • Refer to the SDS for storage, handling, and safety information.
Catalog No. 50-208-1977 Supplier Medchemexpress LLC Supplier No. HY1753110MG
Pyraclonil is a proporphyrinogen oxidase (PPO) inhibitor and herbicide agent supplied as a research-grade solid. The compound has molecular formula C15H15ClN6, molecular weight 314.77 g/mol, and supplier-reported purity of approximately 97%. Intended for laboratory research use; consult the safety data sheet (SDS) for handling and storage instructions.
  • PPO inhibitor suitable for herbicide and biochemical research.
  • High purity (~97%) appropriate for analytical and biological assays.
  • Supplied in a small research pack for method development and screening.
  • Defined molecular weight and formula support accurate dosing.
  • Refer to the SDS for storage, handling, and safety information.