Learn More
Medchemexpress LLC HY-N6626 100mg Medchemexpress, Pyraclostrobin CAS:175013-18-0 Purity:>98%
SDP
Catalog No. 502031560
Encompass_Preferred
Medchemexpress, HY-N6626 100mg Pyraclostrobin CAS:175013-18-0 Pyraclostrobin is a strobilurin fungicide that inhibits mitochondrial complex III of fungal and mammalian cells. Pyraclostrobin induces triglyceride accumulation and triglyceride accumulation in 3T3-L1 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1560 Supplier Medchemexpress LLC Supplier No. HYN6626100MG
Medchemexpress, HY-N6626 100mg Pyraclostrobin CAS:175013-18-0 Pyraclostrobin is a strobilurin fungicide that inhibits mitochondrial complex III of fungal and mammalian cells. Pyraclostrobin induces triglyceride accumulation and triglyceride accumulation in 3T3-L1 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.