Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyraclostrobin | 175013-18-0 | 500 MG
SDP

Catalog No. 5000420037
Encompass_Preferred

Pyraclostrobin is a highly effective, broad-spectrum strobilurin fungicide that can be used to control crop diseases. This off-white to light yellow solid has a purity of 99.91% and is soluble in DMSO. It induces oxidative DNA damage, mitochondrial dysfunction, and autophagy through AMPK/mTOR signaling activation.

  • Highly effective and broad-spectrum strobilurin fungicide
  • Controls crop diseases
  • Induces oxidative DNA damage, mitochondrial dysfunction, and autophagy
  • Purity of 99.91%
  • Off-white to light yellow solid
  • Soluble in DMSO

Catalog No. 50-004-20037 Supplier Medchemexpress LLC Supplier No. HYN6626500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyraclostrobin is a highly effective, broad-spectrum strobilurin fungicide that can be used to control crop diseases. This off-white to light yellow solid has a purity of 99.91% and is soluble in DMSO. It induces oxidative DNA damage, mitochondrial dysfunction, and autophagy through AMPK/mTOR signaling activation.

  • Highly effective and broad-spectrum strobilurin fungicide
  • Controls crop diseases
  • Induces oxidative DNA damage, mitochondrial dysfunction, and autophagy
  • Purity of 99.91%
  • Off-white to light yellow solid
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.