Learn More
Cayman Chemical Pyraclostrobin, Purity: ≥95%, CAS: 175013-18-0, 50 mg
SDP
Catalog No. 502915546
Encompass_Preferred
A carbamate fungicide; active against a variety of fungi (IC50s = 0.18, 0.005, 0.36, 0.03, and 0.15 µg/ml, respectively); is toxic to zebrafish (LC50 = 42.17 µg/L); induces nuclear and mitochondrial deformities in the heart and brain, as well as decreases heart rate, blood volume ejected, and cardiac output, in zebrafish larvae at 37.5 µg/L; decreases locomotor activity and body weight, as well as induces hypothermia and diarrhea, in mice at 400 mg/kg
Catalog No. 50-291-5546 Supplier Cayman Chemical Supplier No. 3753350 MG
A carbamate fungicide; active against a variety of fungi (IC50s = 0.18, 0.005, 0.36, 0.03, and 0.15 µg/ml, respectively); is toxic to zebrafish (LC50 = 42.17 µg/L); induces nuclear and mitochondrial deformities in the heart and brain, as well as decreases heart rate, blood volume ejected, and cardiac output, in zebrafish larvae at 37.5 µg/L; decreases locomotor activity and body weight, as well as induces hypothermia and diarrhea, in mice at 400 mg/kg