Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyraclostrobin-d6 | 99.9% | 393.85 g/mol | C19H12D6ClN3O4 | 1 MG
SDP

Catalog No. 5000284309
Encompass_Preferred

Pyraclostrobin-d6 is the deuterium-labeled isotopologue of the fungicide pyraclostrobin, supplied as a high-purity research standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry, quantitative analysis, and metabolic tracing studies.

  • Deuterium-labeled internal standard for pyraclostrobin.
  • High purity: 99.9%.
  • Chemical formula C19H12D6ClN3O4; molecular weight 393.85 g/mol.
  • Supplied as a 1 mg powder for analytical use.
  • Storage guidance: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-84309 Supplier Medchemexpress LLC Supplier No. HYW6543301MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyraclostrobin-d6 is the deuterium-labeled isotopologue of the fungicide pyraclostrobin, supplied as a high-purity research standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry, quantitative analysis, and metabolic tracing studies.

  • Deuterium-labeled internal standard for pyraclostrobin.
  • High purity: 99.9%.
  • Chemical formula C19H12D6ClN3O4; molecular weight 393.85 g/mol.
  • Supplied as a 1 mg powder for analytical use.
  • Storage guidance: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.