Learn More
Medchemexpress LLC Pyraflufen-ethyl | 129630-19-9 | 98.0% | 25 MG
SDP
Catalog No. 5000340085
Encompass_Preferred
Pyraflufen-ethyl is a phenylpyrazole herbicide for the control of annual and perennial broadleaf weeds. This product is intended for research and analytical applications only.
  • Used for control of annual and perennial broadleaf weeds.
  • Inhibits protoporphyrinogen oxidase (PPO).
  • Controls rosette Parthenium hysterophorus.
  • For research and analytical applications.
Catalog No. 50-003-40085 Supplier Medchemexpress LLC Supplier No. 25MGHY126922
Pyraflufen-ethyl is a phenylpyrazole herbicide for the control of annual and perennial broadleaf weeds. This product is intended for research and analytical applications only.
  • Used for control of annual and perennial broadleaf weeds.
  • Inhibits protoporphyrinogen oxidase (PPO).
  • Controls rosette Parthenium hysterophorus.
  • For research and analytical applications.