Learn More
Medchemexpress LLC Pyraflufen-ethyl | 129630-19-9 | 98.02% | 10 MG
SDP
Catalog No. 5000340847
Encompass_Preferred
Pyraflufen-ethyl is a phenylpyrazole herbicide used for the control of annual and perennial broadleaf weeds. It has a molecular weight of 413.18, a chemical formula of C15H13Cl2F3N2O4, and a CAS No. of 129630-19-9. This product appears as a solid with a color ranging from light yellow to light brown.
  • Used for control of annual and perennial broadleaf weeds
  • Inhibits protoporphyrinogen oxidase (PPO)
  • Controls rosette *Parthenium hysterophorus*
Catalog No. 50-003-40847 Supplier Medchemexpress LLC Supplier No. HY12692210MG
Pyraflufen-ethyl is a phenylpyrazole herbicide used for the control of annual and perennial broadleaf weeds. It has a molecular weight of 413.18, a chemical formula of C15H13Cl2F3N2O4, and a CAS No. of 129630-19-9. This product appears as a solid with a color ranging from light yellow to light brown.
  • Used for control of annual and perennial broadleaf weeds
  • Inhibits protoporphyrinogen oxidase (PPO)
  • Controls rosette *Parthenium hysterophorus*