Learn More
Pyramex™ Alair™ Safety Glasses
Catalog No. 191303478 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Blue Mirror
Clear
Gray
Infinity Blue
Lens Tint:
Blue Mirror
Clear
Gray
Indoor/Outdoor
Infinity Blue
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-3478 Infinity Blue Infinity Blue
19-121-918 Gray Gray
19-121-917 Clear Clear
19-121-919 Blue Mirror Blue Mirror
19-121-920 Clear Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-3478 Supplier Pyramex™ Supplier No. S3260S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Straight temples and integrated nose piece fits all facial sizes.
  • Polycarbonate frame and lens combination are coated for superior scratch resistance
  • Lenses provide 99.9% protection against harmful UV rays
Specifications
Lens Tint Infinity Blue
Frame Color Infinity Blue
Temple Style Straight
Lens Material Polycarbonate