Learn More
MSC Pyramex Cappture Plus Eyewear, Dielectric, H2MAX Clear Lens, 1 ea
Catalog No. 5000676895
Encompass
The Cappture Plus dielectric safety glasses feature H2MAX anti-fog lenses and are designed to comfortably fit over prescription glasses. Their unique temple design avoids interference with prescription frames, ensuring clear vision and comfort.
  • Designed for comprehensive protection over prescription glasses
  • Polycarbonate lenses provide 99% UV protection, lightweight comfort, and a scratch-resistant coating
  • Temples positioned above the frame to prevent interference with prescription lenses
  • Integrated side shields and a flexible rubber gasket offer full eye protection from multi-directional hazards
  • Constructed with non-conductive materials for protection against electrical hazards
  • H2MAX treated lenses ensure maximum clarity in extreme conditions, with abrasion and chemical resistance
Catalog No. 50-006-76895 Supplier MSC Supplier No. 17063124
The Cappture Plus dielectric safety glasses feature H2MAX anti-fog lenses and are designed to comfortably fit over prescription glasses. Their unique temple design avoids interference with prescription frames, ensuring clear vision and comfort.
  • Designed for comprehensive protection over prescription glasses
  • Polycarbonate lenses provide 99% UV protection, lightweight comfort, and a scratch-resistant coating
  • Temples positioned above the frame to prevent interference with prescription lenses
  • Integrated side shields and a flexible rubber gasket offer full eye protection from multi-directional hazards
  • Constructed with non-conductive materials for protection against electrical hazards
  • H2MAX treated lenses ensure maximum clarity in extreme conditions, with abrasion and chemical resistance