Learn More
MSC Pyramex Capstone Shield Goggles, H2X Anti-fog, Clear Lens, 1 ea
Catalog No. 5000675726
Encompass
Capstone® Shield Goggles offer comprehensive eye and face protection. Featuring an H2X Anti-fog lens and soft vinyl body, they fit over prescription glasses and protect against dust, chemical splash. Includes a removable face shield for full coverage and customizable ventilation.
  • Fully adjustable elastic band
  • Fits over prescription glasses
  • H2X Anti-fog coated lens
  • Soft vinyl body for facial contour fit
  • Protects against dust and chemical splash
  • Removable vent caps for ventilation control
  • Integrated removable face shield for full protection
  • Ratchet shield with 3-position adjustment
  • Meets MIL-PRF-32432 High Velocity Impact standards
  • Passes D3 (Droplet/Splash) and D4 (Dust) tests
Catalog No. 50-006-75726 Supplier MSC Supplier No. 81472268
Capstone® Shield Goggles offer comprehensive eye and face protection. Featuring an H2X Anti-fog lens and soft vinyl body, they fit over prescription glasses and protect against dust, chemical splash. Includes a removable face shield for full coverage and customizable ventilation.
  • Fully adjustable elastic band
  • Fits over prescription glasses
  • H2X Anti-fog coated lens
  • Soft vinyl body for facial contour fit
  • Protects against dust and chemical splash
  • Removable vent caps for ventilation control
  • Integrated removable face shield for full protection
  • Ratchet shield with 3-position adjustment
  • Meets MIL-PRF-32432 High Velocity Impact standards
  • Passes D3 (Droplet/Splash) and D4 (Dust) tests