Learn More
Pyramex™ Cortez™ Frame with Navy Temples
Catalog No. 191303521 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
19-130-3521 Clear
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3521 Supplier Pyramex™ Supplier No. SN3610S
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Ribbed, cushioned nosepiece fits a wide range of bridge sizes and protects against slippage.
  • Durable dual injected polycarbonate and rubber temples
  • Single lens design provides maximum protection
  • Lenses are coated for superior scratch resistance
  • Lenses provide 99% protection against harmful UV rays
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Frame Color Navy
Material Polycarbonate, Metal, TPE
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, TPE
UV Protection U6
Frame Type Frameless