Learn More
Pyramex™ Cortez™ Frame Safety Eyewear
Catalog No. 191303509 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Clear lens, Apple temple
Catalog No.
19-130-3509 Clear lens, Apple temple
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3509 Supplier Pyramex™ Supplier No. SA3610S
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Ribbed, cushioned nosepiece fits a wide range of bridge sizes and protects against slippage.
  • Durable dual injected polycarbonate and rubber temples
  • Single lens design provides maximum protection
  • Lenses are coated for superior scratch resistance
  • Lenses provide 99% protection against harmful UV rays
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Lens Material Polycarbonate