Learn More
MSC Pyramex Face Shield, Polycarbonate, Clear, 0.040 in Thick, 1 Each
Catalog No. 5000768852
Encompass
The Pyramex S1020 face shield is constructed from clear polycarbonate, engineered to be fully dielectric with no metal components. It includes a protective peel-away film to maintain clarity until use, ensuring optimal visibility.
  • Clear polycarbonate construction
  • Dimensions: 8 x 15 in, 0.040 in thick
  • Universal pegs for wide headgear compatibility
  • Fully dielectric design with no metal parts
  • Meets ANSI Z87.1+ requirements
  • Includes protective peel-away film
Catalog No. 50-007-68852 Supplier MSC Supplier No. 73375651
The Pyramex S1020 face shield is constructed from clear polycarbonate, engineered to be fully dielectric with no metal components. It includes a protective peel-away film to maintain clarity until use, ensuring optimal visibility.
  • Clear polycarbonate construction
  • Dimensions: 8 x 15 in, 0.040 in thick
  • Universal pegs for wide headgear compatibility
  • Fully dielectric design with no metal parts
  • Meets ANSI Z87.1+ requirements
  • Includes protective peel-away film