Learn More
MSC Pyramex Full Brim Hard Hat, ABS, 4-Point Ratchet, 1 ea
Catalog No. 5000782605
Encompass
The Pyramex HP54160 Ridgeline Full Brim Hard Hat provides robust head protection with an ultra-lightweight design. Constructed from durable ABS material, it features an easy-to-adjust 4-point ratchet suspension system for a secure and comfortable fit, meeting ANSI Z89.1-2014 Type 1, Class E, G, and C standards.
  • Constructed from strong, ultra-light ABS material
  • Ratchet suspension for easy adjustment and customized fit
  • Replaceable soft brow pad for enhanced comfort
  • Suspensions and headbands are replaceable
  • Can be converted to a 6-point suspension system
  • Meets ANSI Z89.1-2014, Type 1, Class E, G, and C safety standards
Catalog No. 50-007-82605 Supplier MSC Supplier No. 17945718
The Pyramex HP54160 Ridgeline Full Brim Hard Hat provides robust head protection with an ultra-lightweight design. Constructed from durable ABS material, it features an easy-to-adjust 4-point ratchet suspension system for a secure and comfortable fit, meeting ANSI Z89.1-2014 Type 1, Class E, G, and C standards.
  • Constructed from strong, ultra-light ABS material
  • Ratchet suspension for easy adjustment and customized fit
  • Replaceable soft brow pad for enhanced comfort
  • Suspensions and headbands are replaceable
  • Can be converted to a 6-point suspension system
  • Meets ANSI Z89.1-2014, Type 1, Class E, G, and C safety standards