Learn More
MSC PYRAMEX Goggles, Anti-Fog, Scratch-Resistant, Lens, Face Shield, 1 ea
Catalog No. 5000687038
Encompass
These safety goggles with an integrated face shield provide full face protection against dust and chemical splash. They feature anti-fog and scratch-resistant clear polycarbonate lenses, offering 99% UVA/B/C protection and enhanced visibility.
  • Adjustable elastic band
  • Fits over prescription glasses
  • Anti-fog coated lenses
  • Soft vinyl goggle body conforms to facial contours
  • Protects against dust and chemical splash
  • Scratch resistant polycarbonate lens offers 99% UVA/B/C protection
  • Removable vent caps for ventilation adjustment or chemical splash resistance
  • Integrated removable shield provides full face protection
  • Ratchet shield allows for 3-position adjustment
  • Polycarbonate shield conforms to face for increased impact and dust protection
Catalog No. 50-006-87038 Supplier MSC Supplier No. 35773027
These safety goggles with an integrated face shield provide full face protection against dust and chemical splash. They feature anti-fog and scratch-resistant clear polycarbonate lenses, offering 99% UVA/B/C protection and enhanced visibility.
  • Adjustable elastic band
  • Fits over prescription glasses
  • Anti-fog coated lenses
  • Soft vinyl goggle body conforms to facial contours
  • Protects against dust and chemical splash
  • Scratch resistant polycarbonate lens offers 99% UVA/B/C protection
  • Removable vent caps for ventilation adjustment or chemical splash resistance
  • Integrated removable shield provides full face protection
  • Ratchet shield allows for 3-position adjustment
  • Polycarbonate shield conforms to face for increased impact and dust protection