Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Hard Hat Adapter

Catalog No. 19151615 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
Blue
Silver
Material:
Aluminum
Nylon
Style:
Dielectric
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Style
19-151-615 Silver Aluminum -
19-151-613 Black Nylon -
19-151-614 Blue Nylon Dielectric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-151-615 Supplier Pyramex™ Supplier No. HHAA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fits most cap-style hard hats and accommodates a variety of faceshields.

  • Universal sizing fits on the brim of most hard hats and accommodates a variety of face shields
  • Note:  Model HHAN (19-151-614) is dielectric

Specifications

Certifications/Compliance ANSI Z87.1
Color Silver
For Use With (Equipment) Cap-style Hard Hats and Variety of Faceshields
Material Aluminum
Product Type Hard Hat Adapter
Size Universal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.