Learn More
Pyramex™ Pyramex™ Hi-Vis T-Shirts
Catalog No. 19555401 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
2X-Large
3X-Large
4X-Large/5X-Large
Large
Medium
X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-401 Hi-vis Orange 4X-Large/5X-Large
19-555-371 Hi-vis Lime Medium
19-555-370 Hi-vis Lime Large
19-555-375 Hi-vis Lime X-Large
19-555-372 Hi-vis Lime 2X-Large
19-555-373 Hi-vis Lime 3X-Large
19-555-374 Hi-vis Lime 4X-Large/5X-Large
19-555-398 Hi-vis Orange Medium
19-555-397 Hi-vis Orange Large
19-555-399 Hi-vis Orange 2X-Large
19-555-400 Hi-vis Orange 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-555-401 Supplier Pyramex™ Supplier No. RTS2120BX4X5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Hi-Vis T-Shirts with ANSI/ISEA 107-2015 safety compliance
  • Hi-Vis lightweight polyester moisture wicking t-shirt with black bottom
  • Outside left chest patch pocket measures 5.5 x 5.5 in.
  • One 2 in. silver reflective stripe applied horizontally across lower chest
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over the shoulders to the top of the back side horizontal stripe
Specifications
Size 4X-Large/5X-Large
Material Polyester
Color Hi-vis Orange
Sleeve Style Short Sleeve
No. of Pockets 1
Certifications/Compliance ANSI/ISEA 107-2015, Type R - Class 2
Size (English) Chest 58 to 64 in.
Tape Type China Brand Silver