Learn More
MSC PYRAMEX Magnifying Safety Glasses, 1.50 Diopter, ANSI Z87.1, 1 ea
Catalog No. 5000769037
Encompass
These safety glasses feature a 1.50 diopter lens with clear, scratch-resistant polycarbonate lenses. They are designed with a full-framed, wrap-around design, black nylon frame, and adjustable bayonet temples, providing reliable UV protection and meeting ANSI Z87.1 and CSA Z94.3-07 standards.
  • Features a 1.50 diopter lens
  • Scratch-resistant coating
  • Polycarbonate lens material
  • Nylon frame material
  • Full-framed, wrap around design
  • Adjustable bayonet temples
  • Provides UV protection
  • Meets CSA Z94.3-07 and ANSI Z87.1 standards
  • Universal size
Catalog No. 50-007-69037 Supplier MSC Supplier No. 80027055
These safety glasses feature a 1.50 diopter lens with clear, scratch-resistant polycarbonate lenses. They are designed with a full-framed, wrap-around design, black nylon frame, and adjustable bayonet temples, providing reliable UV protection and meeting ANSI Z87.1 and CSA Z94.3-07 standards.
  • Features a 1.50 diopter lens
  • Scratch-resistant coating
  • Polycarbonate lens material
  • Nylon frame material
  • Full-framed, wrap around design
  • Adjustable bayonet temples
  • Provides UV protection
  • Meets CSA Z94.3-07 and ANSI Z87.1 standards
  • Universal size