Learn More
MSC PYRAMEX Magnifying Safety Glasses, 2.50 Diopter Amber, ANSI/CSA, 1 ea
Catalog No. 5000787106
Encompass
These Pyramex Venture II series magnifying safety glasses feature a 2.50 diopter lens with amber, scratch-resistant polycarbonate lenses, and comply with ANSI Z87.1 and CSA Z94.3-07 standards for optimal eye protection.
  • Features a 2.50 diopter lens
  • Amber, scratch-resistant polycarbonate lenses
  • Complies with ANSI Z87.1 and CSA Z94.3-07 standards
  • Full-framed, black nylon frame with wrap-around design
  • Bayonet temples with adjustable feature
  • Provides UV protection
  • Nose buds secure the glasses and prevent slipping
  • Universal size
Catalog No. 50-007-87106 Supplier MSC Supplier No. 58177379
These Pyramex Venture II series magnifying safety glasses feature a 2.50 diopter lens with amber, scratch-resistant polycarbonate lenses, and comply with ANSI Z87.1 and CSA Z94.3-07 standards for optimal eye protection.
  • Features a 2.50 diopter lens
  • Amber, scratch-resistant polycarbonate lenses
  • Complies with ANSI Z87.1 and CSA Z94.3-07 standards
  • Full-framed, black nylon frame with wrap-around design
  • Bayonet temples with adjustable feature
  • Provides UV protection
  • Nose buds secure the glasses and prevent slipping
  • Universal size