Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Mini Ztek™ Safety Eyewear

Catalog No. 191300955 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color: black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-0955 Color: black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-0955 Supplier Pyramex™ Supplier No. S2510SN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used for general purposes/indoor applications that require impact protection.

  • Mini Ztek has all the great features of Ztek in a design better suited for slimmer facial sizes.

  • Integrated nose piece. Soft, non-slip rubber temple tips provide non-binding fit.

  • Economical wrap-around single lens provides full panoramic view.

  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection.

Specifications

Lens Coating Impact-resistant
Lens Tint Clear
Product Type Mini Ztek
For Use With (Application) Indoor applications that require impact protection
Frame Color Clear
Lens Material Polycarbonate
UV Protection 99% UVA/B/C protection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.