Learn More
MSC Pyramex Safety Glasses, Frameless, Integrated Nosepiece, ANSI Z87.1, 1 ea
Catalog No. 5000687742
Encompass
Pyramex Ztek safety glasses are frameless eyewear designed for eye protection. They feature an integrated nosepiece and soft, non-slip rubber temple tips for a secure and comfortable fit, suitable for various work environments.
  • Frameless design for wide visibility
  • Integrated nosepiece for comfort
  • Soft, non-slip rubber temple tips for a secure fit
  • Clear lens for general purpose use
  • Meets ANSI/ISEA Z87.1-2020 + High Impact standards
  • Meets CAN/CSA Z94.3-2020 standards
  • Meets CE/UKCA EN 166 standards
Catalog No. 50-006-87742 Supplier MSC Supplier No. 10335909
Pyramex Ztek safety glasses are frameless eyewear designed for eye protection. They feature an integrated nosepiece and soft, non-slip rubber temple tips for a secure and comfortable fit, suitable for various work environments.
  • Frameless design for wide visibility
  • Integrated nosepiece for comfort
  • Soft, non-slip rubber temple tips for a secure fit
  • Clear lens for general purpose use
  • Meets ANSI/ISEA Z87.1-2020 + High Impact standards
  • Meets CAN/CSA Z94.3-2020 standards
  • Meets CE/UKCA EN 166 standards