Learn More
MSC Pyramex Safety Glasses, Frameless, Scratch-Resistant, Gray Polycarbonate Lenses, 1 ea
Catalog No. 5000687692
Encompass
Mini Ztek safety glasses are frameless eyewear designed for slimmer facial sizes, offering comprehensive eye protection with a focus on comfort and durability.
  • Integrated nosepiece
  • Soft, non-slip rubber temple tips for a non-binding fit
  • Scratch-resistant polycarbonate gray lenses
  • UV protection
  • Meets ANSI/ISEA Z87.1-2020 + High Impact standards
  • Wrap-around frame design
  • Non-adjustable temples
Catalog No. 50-006-87692 Supplier MSC Supplier No. 64658446
Mini Ztek safety glasses are frameless eyewear designed for slimmer facial sizes, offering comprehensive eye protection with a focus on comfort and durability.
  • Integrated nosepiece
  • Soft, non-slip rubber temple tips for a non-binding fit
  • Scratch-resistant polycarbonate gray lenses
  • UV protection
  • Meets ANSI/ISEA Z87.1-2020 + High Impact standards
  • Wrap-around frame design
  • Non-adjustable temples