Learn More
MSC Pyramex Safety Glasses, Half-Frame, Gray Lens, ANSI Z87.1-2020, 1 ea
Catalog No. 5000685099
Encompass
PMXTREME safety glasses are designed for comfort and protection, featuring a lightweight, half-frame design with a gray lens and black frame. The panoramic wraparound lens provides an extended field of vision and UV protection, making them suitable for various work environments.
  • All-day comfort from lightweight, half-frame design
  • Enhanced field of vision with panoramic polycarbonate wraparound lens
  • UV protection
  • Comfort grip fit with rubber nose piece and temple tips, ventilated temples
  • Secure strap included
  • Meets ANSI/ISEA Z87.1-2020 standards for eye protection
Catalog No. 50-006-85099 Supplier MSC Supplier No. 73380834
PMXTREME safety glasses are designed for comfort and protection, featuring a lightweight, half-frame design with a gray lens and black frame. The panoramic wraparound lens provides an extended field of vision and UV protection, making them suitable for various work environments.
  • All-day comfort from lightweight, half-frame design
  • Enhanced field of vision with panoramic polycarbonate wraparound lens
  • UV protection
  • Comfort grip fit with rubber nose piece and temple tips, ventilated temples
  • Secure strap included
  • Meets ANSI/ISEA Z87.1-2020 standards for eye protection