Learn More
MSC PYRAMEX Safety Glasses, Scratch-Resistant Polycarbonate, Blue Mirror, Frameless, 1 ea
Catalog No. 5000792977
Encompass
These frameless safety glasses feature scratch-resistant, polycarbonate blue mirror lenses and a black frame. They are part of the Ztek Series, meet ANSI Z87+ standards, and offer UV protection.
  • Frameless design
  • Scratch-resistant lens coating
  • Polycarbonate blue mirror lenses
  • Black frame
  • Non-adjustable temples
  • Ztek series
  • Meets ANSI Z87+ standards
  • UV protection
  • Integrated nose piece
  • Rubber frame material
  • Flexible temples
Catalog No. 50-007-92977 Supplier MSC Supplier No. 55615710
These frameless safety glasses feature scratch-resistant, polycarbonate blue mirror lenses and a black frame. They are part of the Ztek Series, meet ANSI Z87+ standards, and offer UV protection.
  • Frameless design
  • Scratch-resistant lens coating
  • Polycarbonate blue mirror lenses
  • Black frame
  • Non-adjustable temples
  • Ztek series
  • Meets ANSI Z87+ standards
  • UV protection
  • Integrated nose piece
  • Rubber frame material
  • Flexible temples