Learn More
MSC PYRAMEX Safety Glasses, Scratch-Resistant, Polycarbonate, Half-Framed, ANSI Z87.1, 1 ea
Catalog No. 5000685376
Encompass
These safety glasses feature scratch-resistant, blue polycarbonate lenses and a half-framed, wraparound design. The frame is black, and the temples are non-adjustable. They meet ANSI Z87.1+ standards, offering UV protection. The frame material is also polycarbonate.
  • Scratch-resistant lenses
  • Polycarbonate lens material
  • Blue lenses
  • Half-framed, wraparound design
  • Black frame
  • Non-adjustable temples
  • Meets ANSI Z87.1+ standards
  • Provides UV protection
Catalog No. 50-006-85376 Supplier MSC Supplier No. 30298590
These safety glasses feature scratch-resistant, blue polycarbonate lenses and a half-framed, wraparound design. The frame is black, and the temples are non-adjustable. They meet ANSI Z87.1+ standards, offering UV protection. The frame material is also polycarbonate.
  • Scratch-resistant lenses
  • Polycarbonate lens material
  • Blue lenses
  • Half-framed, wraparound design
  • Black frame
  • Non-adjustable temples
  • Meets ANSI Z87.1+ standards
  • Provides UV protection