Learn More
MSC Pyramex Safety Glasses, XS3 Plus, Foam, Anti-fog Clear, 1 ea
Catalog No. 5000769457
Encompass
These XS3 Plus foam padded sealed eyewear feature a unique slim temple design with co-injection temple tips. They include flame resistant foam padding and a detachable elastic strap, passing the D3: Droplet and Splash Test with and without the strap.
  • Unique slim temple design with co-injection temple tips
  • Flame resistant foam padding
  • Detachable elastic strap
  • Passes D3: Droplet and splash test (with and without strap)
  • Custom imprinting available
Catalog No. 50-007-69457 Supplier MSC Supplier No. 52337755
These XS3 Plus foam padded sealed eyewear feature a unique slim temple design with co-injection temple tips. They include flame resistant foam padding and a detachable elastic strap, passing the D3: Droplet and Splash Test with and without the strap.
  • Unique slim temple design with co-injection temple tips
  • Flame resistant foam padding
  • Detachable elastic strap
  • Passes D3: Droplet and splash test (with and without strap)
  • Custom imprinting available