Learn More
MSC PYRAMEX Safety Goggles, Anti-Fog, Scratch-Resistant, Gray Polycarbonate Lenses, 1 ea
Catalog No. 5000675575
Encompass
V2G Safety Goggles offer reliable impact protection for various industrial and laboratory environments. Designed with advanced lens coatings, these goggles enhance visibility and user comfort.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant polycarbonate lenses ensure clear vision
  • Direct vent type for air circulation
  • Impact protection for enhanced safety
  • Gray lens color for glare reduction
  • Black polyvinyl chloride frame for durability
  • Universal size for a comfortable fit
  • UV protection filters harmful rays
  • Meets CSA Z94.3 and ANSI Z87.1+ standards
  • Includes an elastic strap for secure wear
  • Fixed front goggle style
  • Elastic headband material
Catalog No. 50-006-75575 Supplier MSC Supplier No. 82232331
V2G Safety Goggles offer reliable impact protection for various industrial and laboratory environments. Designed with advanced lens coatings, these goggles enhance visibility and user comfort.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant polycarbonate lenses ensure clear vision
  • Direct vent type for air circulation
  • Impact protection for enhanced safety
  • Gray lens color for glare reduction
  • Black polyvinyl chloride frame for durability
  • Universal size for a comfortable fit
  • UV protection filters harmful rays
  • Meets CSA Z94.3 and ANSI Z87.1+ standards
  • Includes an elastic strap for secure wear
  • Fixed front goggle style
  • Elastic headband material