Learn More
MSC Pyramex Safety Goggles, Clear, Anti-Fog, High Impact, 1 ea
Catalog No. 5000765547
Encompass
Pyramex Highlander Safety Goggles are designed for indoor applications requiring impact protection. These goggles feature a dual lens and an inner foam clip for comfort and additional dust protection, ensuring safety and clear vision in various work environments.
  • Lightweight sports styling
  • Dual lens for maximum protection
  • Inner lens clip with soft foam for comfort and extreme styling
  • Wear with or without inner foam for dust protection
  • Optional headband for a goggle fit
  • Exceeds ANSI Z87.1-2010 high impact requirements
  • CE EN166 certified
  • Clear lens for general indoor applications requiring impact protection
  • H2X anti-fog, anti-scratch, anti-static coating
  • H2X coating bonded to lenses for lasting effectiveness
  • H2X coating offers waterproof and excellent scratch resistance
  • H2X passes ANSI Z87.1 and Canadian CAN/CSA Z94.3 standards
Catalog No. 50-007-65547 Supplier MSC Supplier No. 35773209
Pyramex Highlander Safety Goggles are designed for indoor applications requiring impact protection. These goggles feature a dual lens and an inner foam clip for comfort and additional dust protection, ensuring safety and clear vision in various work environments.
  • Lightweight sports styling
  • Dual lens for maximum protection
  • Inner lens clip with soft foam for comfort and extreme styling
  • Wear with or without inner foam for dust protection
  • Optional headband for a goggle fit
  • Exceeds ANSI Z87.1-2010 high impact requirements
  • CE EN166 certified
  • Clear lens for general indoor applications requiring impact protection
  • H2X anti-fog, anti-scratch, anti-static coating
  • H2X coating bonded to lenses for lasting effectiveness
  • H2X coating offers waterproof and excellent scratch resistance
  • H2X passes ANSI Z87.1 and Canadian CAN/CSA Z94.3 standards