Learn More
Pyramex™ Integra™ Safety Glasses
Catalog No. 19051212 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Catalog No. Lens Coating
19-051-212 Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 19-051-212 Supplier Pyramex™ Supplier No. SB410ST
Add to Cart
Edge
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Integra™ Safety Glasses feature single lenses and adjustable temples to ensure reliable performance.
  • Available in a wide variety of frame colors and lens tints
  • Select frames available with fog free coating
  • Lenses are coated for superior scratch resistance
  • Lenses provide 99% protection against harmful UV rays
Specifications
Lens Coating Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Certifications/Compliance Exceed ANSI Z87.1 and CSA Z94.3; CE EN 166 certified
Description Eye protection
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Eye protection
Frame Color Black
Temple Style Adjustable
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection U6
Frame Type Adjustable Temples
Autoclavable Not Autoclavable