Learn More
Pyramex™ V2-Metal™ Safety Glasses
Catalog No. 19121931 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Gray
Frame Color:
Gun Metal
Catalog No. Lens Tint Frame Color
19-121-931 Gray Gun Metal
1 options
This item is not returnable. View return policy
Catalog No. 19-121-931 Supplier Pyramex™ Supplier No. SGM1820S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contemporary metal frame and temples adjust to all facial sizes.
  • Spring loaded temples make this eyewear easy to position
  • 9.75 base curved lens provides full side protection
Specifications
Lens Coating Scratch-resistant
Lens Tint Gray
Product Type Safety Eyewear
For Use With (Application) Outdoor
Frame Color Gun Metal
Temple Style Spring loaded
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Metal
UV Protection 99.9% UVA/UVB/UVC
Frame Type Half Frame