Learn More
Pyramex™ Venture II Readers Safety Glasses
Catalog No. 191303491 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Clear, Gray, Amber, Indoor/Outdoor Mirror
Gray
Mirror, Indoor/Outdoor
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Magnification Power
19-130-3491 Gray +3.00
19-121-921 Clear +1.00
19-121-922 Clear, Gray, Amber, Indoor/Outdoor Mirror +1.50
19-121-923 Clear, Gray, Amber, Indoor/Outdoor Mirror +2.00
19-121-924 Clear, Gray, Amber, Indoor/Outdoor Mirror +2.50
19-130-3481 Clear +3.00
19-130-3483 Mirror, Indoor/Outdoor +1.50
19-130-3484 Mirror, Indoor/Outdoor +2.00
19-130-3485 Mirror, Indoor/Outdoor +2.50
19-130-3489 Gray +2.00
19-130-3490 Gray +2.50
19-130-3488 Gray +1.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-130-3491 Supplier Pyramex™ Supplier No. SB1820R30
Add to Cart
Add to Cart
This item is not returnable. View return policy
Venture II Readers Safety Glasses with base curved lens provide full side protection without distracting seams or traditional type side shields.
  • Nose buds secure the glass and prevent slipping while providing a pillow soft fit
  • Fully adjustable temples - 4 size settings
  • Lenses provide 99% protection against harmful UV rays
  • Lenses are coated for superior scratch resistance
  • Strategic placement of lens insert is out of user's direct line of vision
Specifications
Lens Coating Scratch-resistant, UV-treated, Impact-resistant
Lens Type Magnifying
Lens Tint Gray
Product Type Reader Spectacles
For Use With (Application) For outdoor applications
Frame Color Black
Magnification Power +3.00
Disposable Reusable
Lens Material Polycarbonate
UV Protection U6
Frame Type Half Frame