Learn More
Pyramex™ Zone II™ Safety Eyewear
Catalog No. 19121936 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Blue Wood
Matte Black
Silver
Lens Tint:
Clear
Dark Gray
Gray
Indoor/Outdoor Mirror
Silver
Silver Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-121-936 Silver Silver Mirror
19-121-926 Matte Black Gray
19-121-925 Matte Black Clear
19-121-928 Matte Black Silver
19-121-929 Matte Black Indoor/Outdoor Mirror
19-121-943 Blue Wood Dark Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-121-936 Supplier Pyramex™ Supplier No. SS3370E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Built in nose pads elevate frame for precision fit and weightless wear.
  • Straight back, rubber coated temple tips
  • Lenses filter 99% of harmful UV rays
Specifications
Lens Tint Silver Mirror
Product Type Safety Eyewear
For Use With (Application) Provides protection for eyes
Frame Color Silver
Temple Style Rubber coated tips
Lens Material Polycarbonate