Learn More
Pyramex™ Ztek™ Safety Glasses
Catalog No. 191300953 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Amber
Black
Blue
Clear
Clear Gray
Clear Purple
White
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Blue Mirror
Clear
Coffee
Gold Mirror
Gray
Indoor/Outdoor Mirror
Infinity Blue
Purple Haze
Silver Mirror
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-130-0953 Clear Anti-fog, Scratch-resistant Clear
19-151-315 Black Scratch-resistant Coffee
19-151-318 Blue Scratch-resistant Infinity Blue
19-151-319 Black Scratch-resistant Silver Mirror
19-151-317 Amber Scratch-resistant Amber
19-151-320 Black Scratch-resistant Blue Mirror
19-151-322 Black Scratch-resistant Gold Mirror
19-817-359 Clear Scratch-resistant Indoor/Outdoor Mirror
19-151-316 Black Scratch-resistant Gray
19-817-358 White Scratch-resistant Clear
19-151-703 Clear Purple Scratch-resistant Purple Haze
19-817-420 Clear Gray Anti-fog, Scratch-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-130-0953 Supplier Pyramex™ Supplier No. S2510ST
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Wraparound single lens with panoramic view
  • Lightweight, wraparound frame/lens combination provides superior side protection, without the distraction of seams or bends
  • Soft, rubber temple tips provide comfort without slipping
  • Coated, scratch-resistant lenses provide 99% UV protection
Compliance
Meet ANSI Z87.1 requirements and CSA-Z94.3 standards
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Temple Style Rubber coated tips