Learn More
Medchemexpress LLC HY-D0023 500mg , Pyranine CAS:6358-69-6 Purity:>98%
SDP
Catalog No. 502261154
Encompass_Preferred
HY-D0023 500mg Pyranine CAS: 6358-69-6 Pyranine (HPTS; Solvent Green 7) is a pH-sensitive fluorescent indicator. Pyranine acts as a class of fluorescent chemosensor for the Cu+ ion(λex=450 nm, λem=510 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1154 Supplier Medchemexpress LLC Supplier No. HYD0023500MG
HY-D0023 500mg Pyranine CAS: 6358-69-6 Pyranine (HPTS; Solvent Green 7) is a pH-sensitive fluorescent indicator. Pyranine acts as a class of fluorescent chemosensor for the Cu+ ion(λex=450 nm, λem=510 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.