Learn More
AdipoGen Pyranonigrin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20156M001
Encompass_Preferred
Chemical. CAS: 773855-65-5. Formula: C10H9NO5. MW: 223.2. Isolated from Aspergillus niger FKI-2633. Antioxidant. Radical scavenging activity towards DPPH and superoxide. Shows a suppressive effect on the expression of vascular cell adhesion molecule-1 (VCAM-1) in human umbilical vein endothelial cells (HUVECs) induced by tumor necrosis factor-alpha (TNF-alpha). Insecticidal agent.
Catalog No. 50-205-3541