Learn More
Abcam Pyranonigrin A, Antio x idant agent, 250UG
Catalog No. 502705481
Encompass_Preferred
MW 223.18 Da, Purity >95%. Antioxidant agent. Shows radical scavenging activity toward DPPH and superoxide. Suppresses TNF-α-induced VCAM-1 expression.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5481 Supplier Abcam Supplier No. AB144847250UG
MW 223.18 Da, Purity >95%. Antioxidant agent. Shows radical scavenging activity toward DPPH and superoxide. Suppresses TNF-α-induced VCAM-1 expression.
The product is subject to the following: Abcam Restricted Use Statement