Learn More
U.S. Pharmacopeia Pyrantel Pamoate, 22204-24-6, MFCD00072037, 500mg
Supplier:  U.S. Pharmacopeia 1584003
Encompass
Molecular formula C23H16O6.C11H14N2S, Molecular weight 594.68, Melting Point 476.6 - 509 °F (247 - 265 °C) (decomposes), Synonyms: Pyrantel embonate
Catalog No. NC0119005